2'-Chlor-4'-fluoracetanilid 99%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
N-(2-Chloro-4-fluorophenyl)acetamide
PC1814-250G
PC1814-50G
APOSPC1814-250GEA
48
EUR
APOSPC1814-250G
APOSPC1814-50G
2'-Chlor-4'-fluoracetanilid 99%
2'-Chlor-4'-fluoracetanilid
Formel:
C₈H₇ClFNO Schmelzpunkt: 118 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00042594 CAS-Nummer: 399-35-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...