Kalium(3,5-bis(trifluormethyl)phenyl)trifluoroborat 96%
Lieferant: Apollo Scientific
Synonyme:
Potassium [3,5-bis(trifluoromethyl)phenyl]trifluoroborate, Potassium (3,5-bis(trifluoromethyl)phenyl)trifluoroborate
PC1769-25G
PC1769-5G
APOSPC1769-25GEA
642
EUR
APOSPC1769-25G
APOSPC1769-5G
Kalium(3,5-bis(trifluormethyl)phenyl)trifluoroborat 96%
Kalium(3,5-bis(trifluormethyl)phenyl)trifluoroborat
Formel:
C₈H₃BF₉K Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD03701625 CAS-Nummer: 166328-09-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...