Kalium(4-isobutylphenyl)trifluoroborat 96%
Lieferant: Apollo Scientific
PC1705-1G
PC1705-25G
PC1705-5G
APOSPC1705-1GEA
152
EUR
APOSPC1705-1G
APOSPC1705-25G
APOSPC1705-5G
Kalium(4-isobutylphenyl)trifluoroborat 96%
Kalium(4-isobutylphenyl)trifluoroborat
Formel:
C₁₀H₁₃BF₃K Schmelzpunkt: 290…298 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD04115761 CAS-Nummer: 850623-66-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...