4-Fluor-(2-methoxycarbonyl)phenylboronsäure 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Fluoro-2-methoxycarbonylphenylboronic acid, (4-Fluoro-2-(methoxycarbonyl)phenyl)boronic acid
PC1615-1G
PC1615-250MG
APOSPC1615-1GEA
168
EUR
APOSPC1615-1G
APOSPC1615-250MG
4-Fluor-(2-methoxycarbonyl)phenylboronsäure 95%
4-Fluor-(2-methoxycarbonyl)phenylboronsäure
Formel:
C₈H₈BFO₄ Schmelzpunkt: 86…90 °C Lagerungstemperatur: Kühlschrank |
MDL Number:
MFCD07363762 CAS-Nummer: 871329-81-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...