4-Brom-2-(trifluormethoxy)thiophenol 95+%
Lieferant: Apollo Scientific
|
Achtung
|
PC1593W-10G
PC1593W-1G
APOSPC1593W-10GEA
473
EUR
APOSPC1593W-10G
APOSPC1593W-1G
4-Brom-2-(trifluormethoxy)thiophenol 95+%
4-Brom-2-(trifluormethoxy)thiophenol
Formel:
C₇H₄BrF₃OS Siedepunkt: 96…98 °C/15mmHg Lagerungstemperatur: Kühlschrank |
MDL Number:
MFCD00203474 CAS-Nummer: 175278-15-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...