3,5-Difluoro-4-chlorophenol 97%
Lieferant: Apollo Scientific
Synonyme:
4-Chloro-3,5-difluorophenol
PC1555-10G
PC1555-5G
PC1555-1G
APOSPC1555-10GEA
509
EUR
APOSPC1555-10G
APOSPC1555-5G
APOSPC1555-1G
3,5-Difluoro-4-chlorophenol 97%
3,5-Difluoro-4-chlorophenol
Formel:
C₆H₃ClF₂O Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD06658279 CAS-Nummer: 2268-03-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...