3-Amino-3-(2-fluorphenyl)propionsäure 95%

Lieferant: Apollo Scientific

Synonyme: 3-Amino-3-(2-fluorophenyl)propanoic acid

PC1441-1G PC1441-5G PC1441-10G
APOSPC1441-1GEA 87 EUR
APOSPC1441-1G APOSPC1441-5G APOSPC1441-10G
3-Amino-3-(2-fluorphenyl)propionsäure 95%
3-Amino-3-(2-fluorphenyl)propionsäure
Formel: C₉H₁₀FNO₂
Schmelzpunkt: 222…224 °C
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00193241
CAS-Nummer: 117391-49-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR