Boc-4-(trifluoromethyl)-D-phenylalanine
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-(Trifluoromethyl)-D-phenylalanine, N-BOC protected, (R)-2-((tert-Butoxycarbonyl)amino)-3-(4-(trifluoromethyl)phenyl)propanoic acid
PC1204-1G
PC1204-5G
APOSPC1204-1GEA
106
EUR
APOSPC1204-1G
APOSPC1204-5G
Boc-4-(trifluoromethyl)-D-phenylalanine
Boc-4-(trifluoromethyl)-D-phenylalanine
Formel:
C₁₅H₁₈F₃NO₄ Lagerungstemperatur: Kühlschrank |
MDL Number:
MFCD00797557 CAS-Nummer: 82317-83-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...