Antimon(III)fluorid 98%

Lieferant: Apollo Scientific
Gefahr

Synonyme: Antimony trifluoride

PC1140-1KG PC1140-250G PC1140-50G
APOSPC1140-1KGEA 348 EUR
APOSPC1140-1KG APOSPC1140-250G APOSPC1140-50G
Antimon(III)fluorid 98%
Antimon(III)fluorid

A mild fluorinating agent; Fluorination activity may be increased by addition of Br2, Cl2, or SbCl 5. Swarts, Bull. Acad. Roy. Belg., 1892, 24, 309

Formel: F₃Sb
Siedepunkt: 319 °C
Schmelzpunkt: 292 °C
Dichte: 4.38 g/cm³ (25 °C)
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00011218
CAS-Nummer: 7783-56-4
EINECS: 232-009-2
UN-Nummer: 2923
ADR: 8+6.1,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR