Pentafluorphenyl-2,4,6-trimethylbenzolsulfonat
Lieferant: Apollo Scientific
PC11269-1G
PC11269-500MG
PC11269-5G
APOSPC11269-1GEA
275
EUR
APOSPC11269-1G
APOSPC11269-500MG
APOSPC11269-5G
Pentafluorphenyl-2,4,6-trimethylbenzolsulfonat
Pentafluorphenyl-2,4,6-trimethylbenzolsulfonat
Formel:
C₁₅H₁₁F₅O₃S Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD05975138 CAS-Nummer: 885950-62-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...