Kaliumphenyltrifluoroborat 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: Potassium trifluoro(phenyl)borate, Phenyltrifluoroboric acid potassium salt

PC11221-25G PC11221-5G PC11221-10G
APOSPC11221-25GEA 171 EUR
APOSPC11221-25G APOSPC11221-5G APOSPC11221-10G
Kaliumphenyltrifluoroborat 97%
Kaliumphenyltrifluoroborat
Formel: C₆H₅BF₃K
Schmelzpunkt: 296…301 °C
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD01318172
CAS-Nummer: 153766-81-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR