4-{[2-Fluoro-5-(methylsulphonyl)phenyl]amino}-4-oxobutanoic acid 95+%

Lieferant: Apollo Scientific

PC11200-10G PC11200-1G PC11200-5G
APOSPC11200-10GEA 667 EUR
APOSPC11200-10G APOSPC11200-1G APOSPC11200-5G
4-{[2-Fluoro-5-(methylsulphonyl)phenyl]amino}-4-oxobutanoic acid 95+%
4-{[2-Fluoro-5-(methylsulphonyl)phenyl]amino}-4-oxobutanoic acid
Formel: C₁₁H₁₂FNO₅S
Schmelzpunkt: 176…178 °C
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00203095
CAS-Nummer: 389609-83-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR