2-Chlor-5-(trifluormethyl)phenylboronsäure 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Chloro-5-(trifluoromethyl)phenylboronic Acid, Dihydroxy(2-chloro-5-(trifluoromethyl)phenyl)borane, (2-Chloro-5-(trifluoromethyl)phenyl)metaboric acid, 2-Chloro-5-(trifluoromethyl)benzeneboronic acid, 2-Chloro-5-(trifluoromethyl)phenylboronic Acid (contains varying amounts of Anhydride)

PC11110-25G PC11110-100G
APOSPC11110-25GEA 133 EUR
APOSPC11110-25G APOSPC11110-100G
2-Chlor-5-(trifluormethyl)phenylboronsäure 98%
2-Chlor-5-(trifluormethyl)phenylboronsäure
Formel: C₇H₅BClF₃O₂
Schmelzpunkt: 101…103 °C
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00797335
CAS-Nummer: 182344-18-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR