2-methyl-4-(trifluoromethoxy)phenylacetic acid 98%

Lieferant: Apollo Scientific
Achtung

PC1066-1G PC1066-250MG PC1066-5G
APOSPC1066-1GEA 279 EUR
APOSPC1066-1G APOSPC1066-250MG APOSPC1066-5G
2-methyl-4-(trifluoromethoxy)phenylacetic acid 98%
Formel: C₁₀H₉F₃O₃
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD07368720
CAS-Nummer: 886763-11-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR