3-{4-[(4-Fluorobenzyl)oxy]phenyl}acrylic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-(4-Fluorobenzyloxy)cinnamic acid

PC10584-1G PC10584-250MG
APOSPC10584-1GEA 279 EUR
APOSPC10584-1G APOSPC10584-250MG
3-{4-[(4-Fluorobenzyl)oxy]phenyl}acrylic acid
3-{4-[(4-Fluorobenzyl)oxy]phenyl}acrylic acid
Formel: C₁₆H₁₃FO₃
Schmelzpunkt: 209…210 °C
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00173647
CAS-Nummer: 175136-19-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR