Pentafluorphenyl-2-chlorbenzolsulfonat
Lieferant: Apollo Scientific
PC10566-1G
PC10566-5G
APOSPC10566-1GEA
183
EUR
APOSPC10566-1G
APOSPC10566-5G
Pentafluorphenyl-2-chlorbenzolsulfonat
Pentafluorphenyl-2-chlorbenzolsulfonat
Formel:
C₁₂H₄ClF₅O₃S Schmelzpunkt: 96…97 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD06659541 CAS-Nummer: 885949-55-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...