Pentafluorphenyl-2-naphthalinsulfonat
Lieferant: Apollo Scientific
PC10322-1G
PC10322-500MG
PC10322-5G
APOSPC10322-1GEA
275
EUR
APOSPC10322-1G
APOSPC10322-500MG
APOSPC10322-5G
Pentafluorphenyl-2-naphthalinsulfonat
Pentafluorphenyl-2-naphthalinsulfonat
Formel:
C₁₆H₇F₅O₃S Schmelzpunkt: 105…107 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD05975130 CAS-Nummer: 885950-37-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...