Pentafluorphenyl-4-iodbenzolsulfonat
Lieferant: Apollo Scientific
|
Achtung
|
PC10317-1G
PC10317-5G
APOSPC10317-1GEA
63.8
EUR
APOSPC10317-1G
APOSPC10317-5G
Pentafluorphenyl-4-iodbenzolsulfonat
Pentafluorphenyl-4-iodbenzolsulfonat
Formel:
C₁₂H₄F₅IO₃S Schmelzpunkt: 88…90 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD05975132 CAS-Nummer: 885950-39-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...