Pentafluorphenyl-2,4,6-triisopropylbenzolsulfonat
Lieferant: Apollo Scientific
PC10302-1G
PC10302-500MG
PC10302-5G
APOSPC10302-1GEA
275
EUR
APOSPC10302-1G
APOSPC10302-500MG
APOSPC10302-5G
Pentafluorphenyl-2,4,6-triisopropylbenzolsulfonat
Pentafluorphenyl-2,4,6-triisopropylbenzolsulfonat
Formel:
C₂₁H₂₃F₅O₃S Schmelzpunkt: 42…43 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD05975146 CAS-Nummer: 886361-20-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...