1-Methyl-2-phenethyl-3-(trifluoromethyl)-3-indolinol
Lieferant: Apollo Scientific
PC10255-1G
PC10255-500MG
APOSPC10255-1GEA
1060
EUR
APOSPC10255-1G
APOSPC10255-500MG
1-Methyl-2-phenethyl-3-(trifluoromethyl)-3-indolinol
1-Methyl-2-phenethyl-3-(trifluoromethyl)-3-indolinol
Formel:
C₁₈H₁₈F₃NO Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD03425785 CAS-Nummer: 439095-40-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...