4-[3-(Trifluoromethyl)phenoxy]benzenecarbonitrile
Lieferant: Apollo Scientific
Synonyme:
4-[3-(Trifluoromethyl)phenoxy]benzonitrile
PC10253-1G
PC10253-500MG
PC10253-5G
APOSPC10253-1GEA
210
EUR
APOSPC10253-1G
APOSPC10253-500MG
APOSPC10253-5G
4-[3-(Trifluoromethyl)phenoxy]benzenecarbonitrile
4-[3-(Trifluoromethyl)phenoxy]benzenecarbonitrile
Formel:
C₁₄H₈F₃NO Schmelzpunkt: 59…61 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD01871521 CAS-Nummer: 453557-46-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...