2-(4,4,4-Trifluoro-1-methyl-3-oxobutylidene)-1-hydrazinecarbothioamide

Lieferant: Apollo Scientific

PC10197-1G PC10197-500MG PC10197-5G
APOSPC10197-1GEA 172 EUR
APOSPC10197-1G APOSPC10197-500MG APOSPC10197-5G
2-(4,4,4-Trifluoro-1-methyl-3-oxobutylidene)-1-hydrazinecarbothioamide
2-(4,4,4-Trifluoro-1-methyl-3-oxobutylidene)-1-hydrazinecarbothioamide
Formel: C₆H₈F₃N₃OS
Schmelzpunkt: 144…145 °C
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD03011851
CAS-Nummer: 383148-57-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR