Ethyl 1-(tert-butyl)-5-(trifluoromethyl)-1H-pyrazole-4-carboxylate

Lieferant: Apollo Scientific
Achtung

PC10169-1G PC10169-5G PC10169-250MG
APOSPC10169-1GEA 174 EUR
APOSPC10169-1G APOSPC10169-5G APOSPC10169-250MG
Ethyl 1-(tert-butyl)-5-(trifluoromethyl)-1H-pyrazole-4-carboxylate
Ethyl-1-(tert-butyl)-5-(trifluoromethyl)-1H-pyrazole-4-carboxylate
Formel: C₁₁H₁₅F₃N₂O₂
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD04973716
CAS-Nummer: 852691-03-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR