(3-Fluor-2-methylphenyl)essigsäure 98%

Lieferant: Apollo Scientific

Synonyme: 2-(3-Fluoro-2-methylphenyl)acetic acid

PC0889-1G PC0889-5G PC0889-25G
APOSPC0889-1GEA 43.5 EUR
APOSPC0889-1G APOSPC0889-5G APOSPC0889-25G
(3-Fluor-2-methylphenyl)essigsäure 98%
(3-Fluor-2-methylphenyl)essigsäure
Formel: C₉H₉FO₂
Schmelzpunkt: 90…92 °C
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD03788545
CAS-Nummer: 500912-16-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR