2-Methyl-5-(trifluormethyl)benzonitril 98%
Lieferant: Apollo Scientific
|
Gefahr
|
PC0455-1G
PC0455-250MG
APOSPC0455-1GEA
58
EUR
APOSPC0455-1G
APOSPC0455-250MG
2-Methyl-5-(trifluormethyl)benzonitril 98%
2-Methyl-5-(trifluormethyl)benzonitril
Formel:
C₉H₆F₃N Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD01631594 CAS-Nummer: 261952-03-8 UN-Nummer: 3276 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...