4-Methoxy-3-(trifluormethyl)benzamid

Lieferant: Apollo Scientific

PC0399-10G PC0399-5G
APOSPC0399-10GEA 180 EUR
APOSPC0399-10G APOSPC0399-5G
4-Methoxy-3-(trifluormethyl)benzamid
4-Methoxy-3-(trifluormethyl)benzamid
Formel: C₉H₈F₃NO₂
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD01631493
CAS-Nummer: 261951-86-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR