3-Fluoro-2-(trifluoromethyl)benzyl alcohol 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: (3-Fluoro-2-(trifluoromethyl)phenyl)methanol

PC0393-250MG PC0393-1G PC0393-5G
APOSPC0393-250MGEA 21.9 EUR
APOSPC0393-250MG APOSPC0393-1G APOSPC0393-5G
3-Fluoro-2-(trifluoromethyl)benzyl alcohol 98%
3-Fluoro-2-(trifluoromethyl)benzyl alcohol
Formel: C₈H₆F₄O
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD01631567
CAS-Nummer: 261951-83-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR