2-Chloro-6-fluoro-3-methylphenylacetonitrile 98%

Lieferant: Apollo Scientific
Achtung

PC0123-1G PC0123-5G PC0123-25G
APOSPC0123-1GEA 105 EUR
APOSPC0123-1G APOSPC0123-5G APOSPC0123-25G
2-Chloro-6-fluoro-3-methylphenylacetonitrile 98%
2-Chloro-6-fluoro-3-methylphenylacetonitrile
Formel: C₉H₇ClFN
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD01631364
CAS-Nummer: 261762-94-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR