3-Chlor-2-fluorzimtsäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-(3-Chloro-2-fluorophenyl)acrylic acid
PC0081-10G
PC0081-5G
PC0081-1G
APOSPC0081-10GEA
212
EUR
APOSPC0081-10G
APOSPC0081-5G
APOSPC0081-1G
3-Chlor-2-fluorzimtsäure 98%
3-Chlor-2-fluorzimtsäure
Formel:
C₉H₆ClFO₂ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD01631392 CAS-Nummer: 261762-62-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...