Methyl-4-(isopentyloxy)benzenecarboxylate 95%
Lieferant: Apollo Scientific
OR9867-10G
OR9867-1G
OR9867-5G
APOSOR9867-10GEA
800
EUR
APOSOR9867-10G
APOSOR9867-1G
APOSOR9867-5G
Methyl-4-(isopentyloxy)benzenecarboxylate 95%
Methyl-4-(isopentyloxy)benzenecarboxylate
Formel:
C₁₃H₁₈O₃ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD06659534 CAS-Nummer: 408340-71-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...