(±)-tert-Butyl-2-brombutyrat 98%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
tert-Butyl 2-bromobutanoate, tert-Butyl 2-bromobutyrate
OR965764-100G
OR965764-25G
APOSOR965764-100GEA
481
EUR
APOSOR965764-100G
APOSOR965764-25G
(±)-tert-Butyl-2-brombutyrat 98%
(±)-tert-Butyl-2-brombutyrat
Formel:
C₈H₁₅BrO₂ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00209634 CAS-Nummer: 24457-21-4 UN-Nummer: 1760 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...