Cyclohexylidenecyanoacetic acid 95%

Lieferant: Apollo Scientific

OR965724-1G OR965724-250MG
APOSOR965724-1GEA 517 EUR
APOSOR965724-1G APOSOR965724-250MG
Cyclohexylidenecyanoacetic acid 95%
Formel: C₉H₁₁NO₂
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00013799
CAS-Nummer: 37107-50-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR