2-Chlor-4,5-dimethoxyanilin 95%
Lieferant: Apollo Scientific
OR965602-1G
OR965602-250MG
APOSOR965602-1GEA
719
EUR
APOSOR965602-1G
APOSOR965602-250MG
2-Chlor-4,5-dimethoxyanilin 95%
2-Chlor-4,5-dimethoxyanilin
Formel:
C₈H₁₀ClNO₂ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD08691143 CAS-Nummer: 32829-09-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...