4'-Pentylacetophenon 95%

Lieferant: Apollo Scientific

Synonyme: p-Pentylacetophenone, 4-Pentylacetophenone

OR964868-100G OR964868-25G OR964868-5G
APOSOR964868-100GEA 106 EUR
APOSOR964868-100G APOSOR964868-25G APOSOR964868-5G
4'-Pentylacetophenon 95%
4'-Pentylacetophenon
Formel: C₁₃H₁₈O
Molekulargewicht: 190.29
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00043687
CAS-Nummer: 37593-02-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR