1-Hydroxycyclopentanecarboxylic acid 97%

Lieferant: Apollo Scientific

OR964823-1G OR964823-5G
APOSOR964823-1GEA 34.4 EUR
APOSOR964823-1G APOSOR964823-5G
1-Hydroxycyclopentanecarboxylic acid 97%
1-Hydroxycyclopentanecarboxylic acid
Formel: C₆H₁₀O₃
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00045414
CAS-Nummer: 16841-19-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR