N-Acetyl-D-norvalin 95%
Lieferant: Apollo Scientific
OR964516-1G
OR964516-25G
OR964516-5G
APOSOR964516-1GEA
0
EUR
APOSOR964516-1G
APOSOR964516-25G
APOSOR964516-5G
N-Acetyl-D-norvalin 95%
N-Acetyl-D-norvalin
Formel:
C₇H₁₃NO₃ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00151962 CAS-Nummer: 57357-56-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...