N-α-Fmoc-N-g-tosyl-D-arginine 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Fmoc-D-Arg(Tos)-OH
OR963991-1G
OR963991-5G
APOSOR963991-1GEA
183
EUR
APOSOR963991-1G
APOSOR963991-5G
N-α-Fmoc-N-g-tosyl-D-arginine 98%
N-alpha-Fmoc-N-g-tosyl-D-arginine
Formel:
C₂₈H₃₀N₄O₆S Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00065621 CAS-Nummer: 139090-50-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...