6-Methoxy-2-naphthoesäure
Lieferant: Apollo Scientific
|
Achtung
|
OR9637-1G
OR9637-5G
APOSOR9637-1GEA
21.9
EUR
APOSOR9637-1G
APOSOR9637-5G
6-Methoxy-2-naphthoesäure
6-Methoxy-2-naphthoesäure
Formel:
C₁₂H₁₀O₃ Schmelzpunkt: 192…199 °C Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00092750 CAS-Nummer: 2471-70-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...