Methyl-2-aminohexanoate hydrochloride 98%

Lieferant: Apollo Scientific

Synonyme: H-DL-NorLeu-OMe·HCl, H-DL-Nle-OMe·HCl, Methyl 2-aminohexanoate hydrochloride

OR963597-100G OR963597-25G
APOSOR963597-100GEA 400 EUR
APOSOR963597-100G APOSOR963597-25G
Methyl-2-aminohexanoate hydrochloride 98%
Methyl-2-aminohexanoate hydrochloride
Formel: C₇H₁₆ClNO₂
Molekulargewicht: 181.66 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00058296
CAS-Nummer: 77300-48-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR