Fmoc-Cys(Me)-OH

Lieferant: Apollo Scientific
Achtung

Synonyme: N-Fmoc-S-methyl-L-cysteine

OR963134-1G OR963134-5G
APOSOR963134-1GEA 21.9 EUR
APOSOR963134-1G APOSOR963134-5G
Fmoc-Cys(Me)-OH
Fmoc-Cys(Me)-OH
Formel: C₁₉H₁₉NO₄S
Molekulargewicht: 357.424 g/mol
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD00237014
CAS-Nummer: 138021-87-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR