Fmoc-N-Me-D-Leu-OH 97%
Lieferant: Apollo Scientific
Synonyme:
N-Fmoc-N-methyl-D-leucine, (R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)(methyl)amino)-4-methylpentanoic acid, Fmoc-D-N-Me-Leu-OH
OR963073-25G
OR963073-5G
APOSOR963073-25GEA
394
EUR
APOSOR963073-25G
APOSOR963073-5G
Fmoc-N-Me-D-Leu-OH 97%
Fmoc-N-Me-D-Leu-OH
Formel:
C₂₂H₂₅NO₄ Molekulargewicht: 367.445 g/mol Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00235877 CAS-Nummer: 103478-63-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...