4-(2-Methyl-1H-imidazol-1-yl)benzaldehyde 97%
Lieferant: Apollo Scientific
OR962633-1G
OR962633-5G
APOSOR962633-1GEA
46.4
EUR
APOSOR962633-1G
APOSOR962633-5G
4-(2-Methyl-1H-imidazol-1-yl)benzaldehyde 97%
4-(2-Methyl-1H-imidazol-1-yl)benzaldehyde
Formel:
C₁₁H₁₀N₂O Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD05864665 CAS-Nummer: 88427-96-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...