Boc-N-ME-DL-PHG-OH 97%
Lieferant: Apollo Scientific
Synonyme:
N-Boc-a-(methylamino)benzeneacetic acid
OR962189-1G
OR962189-25G
OR962189-5G
APOSOR962189-1GEA
139
EUR
APOSOR962189-1G
APOSOR962189-25G
APOSOR962189-5G
Boc-N-ME-DL-PHG-OH 97%
Boc-N-ME-DL-PHG-OH
Formel:
C₁₄H₁₉NO₄ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD06796751 CAS-Nummer: 30925-08-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...