(R)-3-(Fmoc-amino)-3-phenylpropionsäure 97%
Lieferant: Apollo Scientific
Synonyme:
(R)-3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-phenylpropanoic acid
OR962068-1G
OR962068-5G
APOSOR962068-1GEA
79.9
EUR
APOSOR962068-1G
APOSOR962068-5G
(R)-3-(Fmoc-amino)-3-phenylpropionsäure 97%
(R)-3-(Fmoc-amino)-3-phenylpropionsäure
Formel:
C₂₄H₂₁NO₄ Molekulargewicht: 387.43 g/mol Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD01311783 CAS-Nummer: 220498-02-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...