Boc-n-Me-abz-OH 97%

Lieferant: Apollo Scientific

Synonyme: 2-((tert-Butoxycarbonyl)(methyl)amino)benzoic acid, 2-(N-Boc-N-methylamino)benzoic acid

OR960977-1G OR960977-5G
APOSOR960977-1GEA 163 EUR
APOSOR960977-1G APOSOR960977-5G
Boc-n-Me-abz-OH 97%
Boc-n-Me-abz-OH
Formel: C₁₃H₁₇NO₄
Molekulargewicht: 251.282 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00237567
CAS-Nummer: 141871-02-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR