(S)-Methyl-2-aminohexanoate hydrochloride 98%

Lieferant: Apollo Scientific

Synonyme: H-Nle-OMe·HCl, (S)-Methyl 2-aminohexanoate hydrochloride, L-Norleucine methyl ester hydrochloride

OR960947-100G OR960947-25G OR960947-5G
APOSOR960947-100GEA 403 EUR
APOSOR960947-100G APOSOR960947-25G APOSOR960947-5G
(S)-Methyl-2-aminohexanoate hydrochloride 98%
(S)-Methyl-2-aminohexanoate hydrochloride
Formel: C₇H₁₆ClNO₂
Molekulargewicht: 181.66 g/mol
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD00077151
CAS-Nummer: 3844-54-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR