2'-Deoxyguanosine monohydrate 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Amino-9-((2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-1,9-dihydro-6H-purin-6-one hydrate
OR960833-100G
OR960833-25G
OR960833-5G
APOSOR960833-100GEA
475
EUR
APOSOR960833-100G
APOSOR960833-25G
APOSOR960833-5G
2'-Deoxyguanosine monohydrate 98%
2'-Deoxyguanosine monohydrate
Formel:
C₁₀H₁₃N₅O₄·H₂O Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00150760 CAS-Nummer: 312693-72-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...