Boc-ARG(PMC)-OH 99%

Lieferant: Apollo Scientific
Achtung

OR960778-1G OR960778-5G
APOSOR960778-1GEA 176 EUR
APOSOR960778-1G APOSOR960778-5G
Boc-ARG(PMC)-OH 99%
Boc-ARG(PMC)-OH
Formel: C₂₅H₄₀N₄O₇S
Molekulargewicht: 540.676 g/mol
Lagerungstemperatur: Kühlschrank
MDL Number: MFCD00236823
CAS-Nummer: 200125-12-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR