1-Ethyl-2-nitrobenzol 99%
Lieferant: Apollo Scientific
Synonyme:
2-Ethylnitrobenzene
OR956256-100G
OR956256-1KG
OR956256-500G
OR956256-25G
APOSOR956256-100GEA
48
EUR
APOSOR956256-100G
APOSOR956256-1KG
APOSOR956256-500G
APOSOR956256-25G
1-Ethyl-2-nitrobenzol 99%
1-Ethyl-2-nitrobenzol
Formel:
C₈H₉NO₂ Lagerungstemperatur: Raumtemperatur |
MDL Number:
MFCD00024294 CAS-Nummer: 612-22-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...