H-Phg(4-Cl)-OH 97%

Lieferant: Apollo Scientific

Synonyme: H-Phg(4-Cl)-OH, 4-Chloro-L-2-phenylglycine, L-2-(4-Chlorophenyl)glycine, (S)-2-Amino-2-(4-chlorophenyl)acetic acid

OR953851-25G OR953851-5G
APOSOR953851-25GEA 265 EUR
APOSOR953851-25G APOSOR953851-5G
H-Phg(4-Cl)-OH 97%
H-Phg(4-Cl)-OH
Formel: C₈H₈ClNO₂
Lagerungstemperatur: Raumtemperatur
MDL Number: MFCD03701467
CAS-Nummer: 67336-19-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR